Learn More
Medchemexpress LLC Uridine triphosphate | 00-00-0 | MFCD00274571 | 98.0% | 497.99 g/mol | C9H13Li2(15N)2O15P3 | 5 MG
SDP
Catalog No. 5000240712
Encompass_Preferred
Uridine triphosphate-15N2 dilithium is a stable isotope-labeled uridine 5′-triphosphate supplied as the dilithium salt for biochemical and analytical research. It functions as a substrate for enzymatic RNA synthesis, a tracer for metabolic studies, and an internal standard for mass spectrometry, and is provided in small research-scale quantities.
  • Purity of 98.0%.
  • 15N2 isotope label for metabolic tracing and mass spectrometry applications.
  • Dilithium salt form for improved solubility and stability in aqueous buffers.
  • Suitable as a substrate for RNA synthesis reactions.
  • Available in small research sizes such as 1 mg, 5 mg, and 10 mg.
Catalog No. 50-002-40712 Supplier Medchemexpress LLC Supplier No. HY107372S45MG
Uridine triphosphate-15N2 dilithium is a stable isotope-labeled uridine 5′-triphosphate supplied as the dilithium salt for biochemical and analytical research. It functions as a substrate for enzymatic RNA synthesis, a tracer for metabolic studies, and an internal standard for mass spectrometry, and is provided in small research-scale quantities.
  • Purity of 98.0%.
  • 15N2 isotope label for metabolic tracing and mass spectrometry applications.
  • Dilithium salt form for improved solubility and stability in aqueous buffers.
  • Suitable as a substrate for RNA synthesis reactions.
  • Available in small research sizes such as 1 mg, 5 mg, and 10 mg.