Learn More
Medchemexpress LLC Uridine triphosphate-13C9 dilithium | 00-00-0 | 98.0% | 504.94 g/mol | 13C9H13Li2N2O15P3 | 5 MG
SDP
Catalog No. 5000240864
Encompass_Preferred
Uridine triphosphate-13C9 dilithium is an isotopically labeled uridine 5'-triphosphate supplied as the dilithium salt for use as an internal standard and tracer in biochemical and analytical workflows, including mass spectrometry and nucleotide metabolism studies.
  • Isotopically labeled with 13C9 for accurate mass-based quantification.
  • Supplied as the dilithium salt for improved solubility and stability.
  • High purity suitable for analytical applications.
  • Available in small vials for trace-level experiments.
  • Includes data sheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-40864 Supplier Medchemexpress LLC Supplier No. HY107372S25MG
Uridine triphosphate-13C9 dilithium is an isotopically labeled uridine 5'-triphosphate supplied as the dilithium salt for use as an internal standard and tracer in biochemical and analytical workflows, including mass spectrometry and nucleotide metabolism studies.
  • Isotopically labeled with 13C9 for accurate mass-based quantification.
  • Supplied as the dilithium salt for improved solubility and stability.
  • High purity suitable for analytical applications.
  • Available in small vials for trace-level experiments.
  • Includes data sheet, certificate of analysis, and safety data sheet.