Learn More
Medchemexpress LLC Uridine triphosphate-15N2 dilithium | 00-00-0 | 98.0% | 497.99 | C9H13Li215N2O15P3 | 10 MG
SDP
Catalog No. 5000241384
Encompass_Preferred
Uridine triphosphate-15N2 dilithium is an isotopically labeled form of uridine 5'-triphosphate supplied as the dilithium salt for research use. It is formulated as a concentrated solution suitable for biochemical, analytical, and mass spectrometry applications that require stable isotope-labeled nucleotide standards or tracers.
  • 15N2 isotopically labeled uridine triphosphate for tracing and mass spectrometry
  • Purity 98.0% ensures reliable analytical performance
  • Supplied as a dilithium salt in a liquid, colorless to light yellow form
  • Packaged as 10 mg with 100 mM concentration in Tris·HCl buffer (pH 7.5)
  • Intended for research use only
Catalog No. 50-002-41384 Supplier Medchemexpress LLC Supplier No. HY107372S410MG
Uridine triphosphate-15N2 dilithium is an isotopically labeled form of uridine 5'-triphosphate supplied as the dilithium salt for research use. It is formulated as a concentrated solution suitable for biochemical, analytical, and mass spectrometry applications that require stable isotope-labeled nucleotide standards or tracers.
  • 15N2 isotopically labeled uridine triphosphate for tracing and mass spectrometry
  • Purity 98.0% ensures reliable analytical performance
  • Supplied as a dilithium salt in a liquid, colorless to light yellow form
  • Packaged as 10 mg with 100 mM concentration in Tris·HCl buffer (pH 7.5)
  • Intended for research use only