Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uridine triphosphate trisodium dihydrate | 116295-90-0 | MFCD00150714 | 97.0% | 586.12 g/mol | C9H16N2Na3O17P3 | 500 MG
SDP

Catalog No. 5000241528
Encompass_Preferred

Uridine triphosphate trisodium dihydrate is the trisodium salt dihydrate form of UTP used as a nucleotide triphosphate donor in biochemical and molecular biology applications. It is supplied as a white to off-white solid and is commonly used in enzymatic assays, RNA synthesis, and other nucleotide-dependent reactions.

  • High purity (97.0%).
  • Chemical formula C9H16N2Na3O17P3; molecular weight 586.12 g/mol.
  • White to off-white solid, convenient for weighing and dissolution.
  • Suitable as a nucleotide triphosphate donor in enzymatic assays and RNA synthesis.
  • Store sealed at -20°C and protect from moisture.

Catalog No. 50-002-41528 Supplier Medchemexpress LLC Supplier No. HY107372A500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uridine triphosphate trisodium dihydrate is the trisodium salt dihydrate form of UTP used as a nucleotide triphosphate donor in biochemical and molecular biology applications. It is supplied as a white to off-white solid and is commonly used in enzymatic assays, RNA synthesis, and other nucleotide-dependent reactions.

  • High purity (97.0%).
  • Chemical formula C9H16N2Na3O17P3; molecular weight 586.12 g/mol.
  • White to off-white solid, convenient for weighing and dissolution.
  • Suitable as a nucleotide triphosphate donor in enzymatic assays and RNA synthesis.
  • Store sealed at -20°C and protect from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.