Learn More
Medchemexpress LLC Uridine triphosphate trisodium salt | 19817-92-6 | 550.09 | 1 G
SDP
Catalog No. 5000454188
Encompass_Preferred
Uridine triphosphate (trisodium salt) (Standard) is the analytical standard of Uridine triphosphate (trisodium salt). This product is intended for research and analytical applications. Uridine triphosphate (UTP) trisodium salt is a pyrimidine nucleoside triphosphate that is used as a substrate to synthesize RNA or as an energy source in metabolic reactions. Uridine triphosphate trisodium salt activates membrane-bound P2Y2 receptors.
  • Analytical standard
  • Intended for research and analytical applications
  • Pyrimidine nucleoside triphosphate
  • Used as a substrate to synthesize RNA
  • Used as an energy source in metabolic reactions
  • Activates membrane-bound P2Y2 receptors
Catalog No. 50-004-54188 Supplier Medchemexpress LLC Supplier No. HYW0130931G
Uridine triphosphate (trisodium salt) (Standard) is the analytical standard of Uridine triphosphate (trisodium salt). This product is intended for research and analytical applications. Uridine triphosphate (UTP) trisodium salt is a pyrimidine nucleoside triphosphate that is used as a substrate to synthesize RNA or as an energy source in metabolic reactions. Uridine triphosphate trisodium salt activates membrane-bound P2Y2 receptors.
  • Analytical standard
  • Intended for research and analytical applications
  • Pyrimidine nucleoside triphosphate
  • Used as a substrate to synthesize RNA
  • Used as an energy source in metabolic reactions
  • Activates membrane-bound P2Y2 receptors