Learn More
Medchemexpress LLC Uridine triphosphate trisodium salt | 19817-92-6 | 97.3% | 550.09 | 1 ML
SDP
Catalog No. 5000454616
Encompass_Preferred
Uridine triphosphate (UTP) trisodium salt is a pyrimidine nucleoside triphosphate that is used as a substrate to synthesize RNA or as an energy source in metabolic reactions. It also activates membrane-bound P2Y2 receptors.
  • Pyrimidine nucleoside triphosphate
  • Used as a substrate to synthesize RNA
  • Functions as an energy source in metabolic reactions
  • Activates membrane-bound P2Y2 receptors
  • Enhances RT4-D6P2T cells migration and wound repair
  • Induces Schwannoma cell migration through activation of P2Y2 receptors and by increasing extracellular MMP-2 activation and expression
  • Evokes MAPK phosphorylation in a biphasic pattern
  • Reduces infarct size and improves rat heart function after myocardial infarct
Catalog No. 50-004-54616 Supplier Medchemexpress LLC Supplier No. HYW01309310MM/1ML
Uridine triphosphate (UTP) trisodium salt is a pyrimidine nucleoside triphosphate that is used as a substrate to synthesize RNA or as an energy source in metabolic reactions. It also activates membrane-bound P2Y2 receptors.
  • Pyrimidine nucleoside triphosphate
  • Used as a substrate to synthesize RNA
  • Functions as an energy source in metabolic reactions
  • Activates membrane-bound P2Y2 receptors
  • Enhances RT4-D6P2T cells migration and wound repair
  • Induces Schwannoma cell migration through activation of P2Y2 receptors and by increasing extracellular MMP-2 activation and expression
  • Evokes MAPK phosphorylation in a biphasic pattern
  • Reduces infarct size and improves rat heart function after myocardial infarct