Learn More
Medchemexpress LLC Uridine triphosphate trisodium salt | 19817-92-6 | 97.3% | 550.09 | 5 G
SDP
Catalog No. 5000454436
Encompass_Preferred
Uridine triphosphate (UTP) trisodium salt is a pyrimidine nucleoside triphosphate that is used as a substrate to synthesize RNA or as an energy source in metabolic reactions. It also activates membrane-bound P2Y2 receptors.
  • Used as a substrate to synthesize RNA
  • Functions as an energy source in metabolic reactions
  • Activates membrane-bound P2Y2 receptors
  • Enhances RT4-D6P2T cells migration and wound repair in vitro
  • Induces Schwannoma cell migration through activation of P2Y2 receptors and increased extracellular MMP-2 activation and expression in vitro
  • Evokes MAPK phosphorylation in a biphasic pattern in vitro
  • Reduces infarct size and improves rat heart function after myocardial infarct in vivo
Catalog No. 50-004-54436 Supplier Medchemexpress LLC Supplier No. HYW0130935G
Uridine triphosphate (UTP) trisodium salt is a pyrimidine nucleoside triphosphate that is used as a substrate to synthesize RNA or as an energy source in metabolic reactions. It also activates membrane-bound P2Y2 receptors.
  • Used as a substrate to synthesize RNA
  • Functions as an energy source in metabolic reactions
  • Activates membrane-bound P2Y2 receptors
  • Enhances RT4-D6P2T cells migration and wound repair in vitro
  • Induces Schwannoma cell migration through activation of P2Y2 receptors and increased extracellular MMP-2 activation and expression in vitro
  • Evokes MAPK phosphorylation in a biphasic pattern in vitro
  • Reduces infarct size and improves rat heart function after myocardial infarct in vivo