Learn More
Medchemexpress LLC Uridine triphosphate trisodium salt | 19817-92-6 | 97.3% | 550.09 | 500 MG
SDP
Catalog No. 5000454334
Encompass_Preferred
Uridine triphosphate trisodium salt is a pyrimidine nucleoside triphosphate that is used as a substrate to synthesize RNA or as an energy source in metabolic reactions. It also activates membrane-bound P2Y2 receptors.
  • Used as a substrate to synthesize RNA or as an energy source in metabolic reactions.
  • Activates membrane-bound P2Y2 receptors.
  • Enhances RT4-D6P2T cells migration and wound repair in vitro.
  • Induces Schwannoma cell migration through activation of P2Y2 receptors and increased extracellular MMP-2 activation and expression in vitro.
  • Evokes MAPK phosphorylation in a biphasic pattern in vitro.
  • Reduces infarct size and improves rat heart function after myocardial infarct in vivo.
Catalog No. 50-004-54334 Supplier Medchemexpress LLC Supplier No. HYW013093500MG
Uridine triphosphate trisodium salt is a pyrimidine nucleoside triphosphate that is used as a substrate to synthesize RNA or as an energy source in metabolic reactions. It also activates membrane-bound P2Y2 receptors.
  • Used as a substrate to synthesize RNA or as an energy source in metabolic reactions.
  • Activates membrane-bound P2Y2 receptors.
  • Enhances RT4-D6P2T cells migration and wound repair in vitro.
  • Induces Schwannoma cell migration through activation of P2Y2 receptors and increased extracellular MMP-2 activation and expression in vitro.
  • Evokes MAPK phosphorylation in a biphasic pattern in vitro.
  • Reduces infarct size and improves rat heart function after myocardial infarct in vivo.