Learn More
Labcon North America UrineTime™ II 24 Hour Urine Collection System, Case of 4
Catalog No. NC1514672
Encompass_Preferred
Container; Urine; 24 Hour; Labcon; UrineTime™ II; 24 Hour urine collection and proportioning system; Automatically splits urine samples into two equal containers; Allows for faster diagnosis; Perfect for trials and other clinical use; Includes two polyethylene 1 liter containers, funnel, plugs, instructions, and disposal bags ; Not Autoclavable; Capacity: 1L; 4 assembled kits per case; Made in USA
Catalog No. NC1514672 Supplier Labcon North America Supplier No. 40104200109
Container; Urine; 24 Hour; Labcon; UrineTime™ II; 24 Hour urine collection and proportioning system; Automatically splits urine samples into two equal containers; Allows for faster diagnosis; Perfect for trials and other clinical use; Includes two polyethylene 1 liter containers, funnel, plugs, instructions, and disposal bags ; Not Autoclavable; Capacity: 1L; 4 assembled kits per case; Made in USA