Description
Advanced gout kit.
- Includes polarizer, analyzer and compensator
- Slide in easily in the available slot under the objective turret
- Recommended for gout fluid analysis
- Combine with phase contrast for urinalysis applications
Specifications
Specifications
| Type | Adapter Kit |
| Autoclavable | Not Autoclavable |
| For Use With (Equipment) | SEBAP4C2 Microscope |
| Sterility | Sterile |
| Voltage | 100/115/220/230 V |