Learn More
Ascensia Diabetes Care Urinalysis Reagent Test Strips
Catalog No. AM2883
Change view
Click to view available options
Detectable Analytes:
Glucose
Glucose, Ketone
Quantity:
100 Pack
50 Pack
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes Quantity
AM-2883 Glucose, Ketone 50 Pack
AM-2803 Glucose 100 Pack
AM-2882 Glucose, Ketone 100 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AM-2883 Supplier Ascensia Diabetes Care Supplier No. 7081683
Add to Cart
Add to Cart
This item is not returnable. View return policy
Single-use, plastic strips with multiple reagent test areas
  • Ready to use and entirely disposable
  • Dip and read visually for results
  • Some strips can be read instrumentally on Clinitek™ 50, Clinitek 100, Clinitek 200, Clinitek 200+ and Clinitek 500 Urine Chemistry analyzers
Specifications
Clia Complexity Waived
Detectable Analytes Glucose, Ketone
Disposable Single-use
For Use With (Application) Urinalysis
Quantity 50 Pack