Learn More
3B Scientific™ Urinalysis Using Simulated Urine
SDPSCI_ED
Catalog No. S00214 Shop All 3B Scientific Products
Change view
Click to view available options
Size:
Materials for 15 Groups of Students
Catalog No. Size
S00214 Materials for 15 Groups of Students
1 options
This item is not returnable. View return policy
Catalog No. S00214 Supplier 3B Scientific™ Supplier No. 3007473
This item is not returnable. View return policy
In this urinalysis activity, students will use simulated urine to avoid the unpleasantness of using the real thing while still performing actual tests used on real urine samples. Kit contains enough materials for 15 groups of students.
  • Kit Includes:
    • 4 Simulated Urine Samples, 250 mL Each
      • Patient X
      • Patient Y
      • Patient Z
      • Control
    • 2 Bottles of Benedict's Reagent, 25 mL
    • 2 Bottles of Biuret Solution, 25 mL
    • 2 Bottles of Sulkowitch's Reagent, 25 mL
    • 1 Package of pH Test Strips, 100/vial
    • 1 Box Microscope Slides
    • 1 Package of Coverslips
    • 60 Graduated Plastic Cups, 30 mL
    • 60 Graduated Pipettes, 1 mL
Specifications
Product Type Urinalysis Simulation Kit
For Use With (Equipment) Hot Water Bath, Glass Test Tubes and Compound Microscopes
Includes 4 Simulated Urine Samples, 2 Bottles of Benedict's Reagent, 2 Bottles of Biuret Solution, 2 Bottles of Sulkowitch's Reagent, 1 Package of pH Test Strips, 1 Box Microscope Slides, 1 Package of Coverslips, 60 Graduated Plastic Cups, 60 Graduated Pipettes
Size Materials for 15 Groups of Students