Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ImmuView™ S. pneumoniae and L. pneumophila Urinary Antigen Test

Catalog No. 23900563
Change view
Click to view available options
Format:
Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
23-900-563 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-900-563 Supplier ImmuView™ Supplier No. 10557
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An in vitro rapid lateral flow test intended for the qualitative detection of Streptococcus pneumoniae and Legionella pneumophila antigens in urine specimens from patients with symptoms of pneumonia.

  • The assay is further intended to aid in the diagnosis of S. pneumoniae infections by detection of S. pneumoniae antigen in cerebrospinal fluid (CSF).
  • 2 results with 1 test
  • Similar symptoms, different treatment
  • Detects Legionellaserogroups 1, 3, 6, 8, 10, and 12

Result Indicators

  • The control line appears at top of the strip.
  • A positive result for pneumophila will produce a blue line in the middle of the strip.
  • A positive result for S. pneumoniae will produce a pink/red line at the bottom of the strip.

Specifications

CE Marker CE-IVD
Detectable Analytes S. pneumoniae, L. pneumophila
Format Kit
For Use With (Application) Lateral Flow Immunoassay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.