Learn More
Oxford Biomedical Research Inc URINARY ISOPROSTANE ELISA KIT
SDP
Supplier:  Oxford Biomedical Research Inc EA85
Encompass
Kit, Urinary Isoprostane Assay; There is strong evidence that isoprostanes are excellent molecular biomarkers for oxidant stress; 15-F2t-Isoprostane (previously called 8-epi-PGF2a) recognized as the gold standard of oxidant
Catalog No. NC9762048