Learn More
Crystal Chem Inc Urinary (NAG) Assay Kit, 96-tests
SDP
Catalog No. 501947675
Encompass_Preferred
The Urinary (NAG) Assay is a highly sensitive assay that is used to quantify levels of NAG in human urine in less than 10 minutes. Highlights: Kit uses sample size of only µL of urine. Stable liquid stable format requires no reagent preparation.Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7675 Supplier Crystal Chem Inc Supplier No. 80219
The Urinary (NAG) Assay is a highly sensitive assay that is used to quantify levels of NAG in human urine in less than 10 minutes. Highlights: Kit uses sample size of only µL of urine. Stable liquid stable format requires no reagent preparation.Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.