Learn More
Creative Biomart Urinary Trypsin Inhibitor (UTI)
Catalog No. 502084269
Encompass_Preferred
Recombinant Ulinastatin produced in Yeast is a single, glycosylated, polypeptide having 143 amino acids, molecular mass of 20 kDa and predicted pI of 4.5.
Catalog No. 50-208-4269 Supplier Creative Biomart Supplier No. THP0218
Recombinant Ulinastatin produced in Yeast is a single, glycosylated, polypeptide having 143 amino acids, molecular mass of 20 kDa and predicted pI of 4.5.