Learn More
Onsite Testing Specialists Inc URINE ADULTERANT 25PK/INTECT#7
Catalog No. NC9071249
Encompass_Preferred
Test for Creatinine, Nitrite, Glutaraldehyde, pH, Specific Gravity, Bleach & Pyridinium Chlorochromate
Catalog No. NC9071249 Supplier Onsite Testing Specialists Inc Supplier No. AD007
Add to Cart
Add to Cart
Test for Creatinine, Nitrite, Glutaraldehyde, pH, Specific Gravity, Bleach & Pyridinium Chlorochromate