Learn More
JANT PHARMACAL CORPORATION ACCUTEST Urine Adulteration Detection Strips Creatinine, Nitrite, Glutaraldehyde, pH, Specific Gravity, Bleach, Pyridinium Chlorochromate) 25 Tests/Kit
SDP
Catalog No. NC3457119
Encompass_Preferred
A rapid screening test for the simultaneous detection of Creatinine, Nitrite, Glutaraldehyde, pH, Specific Gravity, and Oxidants/Pyridinium Chlorochromate (PCC) in human urine.
Catalog No. NC3457119 Supplier JANT PHARMACAL CORPORATION Supplier No. DS78
A rapid screening test for the simultaneous detection of Creatinine, Nitrite, Glutaraldehyde, pH, Specific Gravity, and Oxidants/Pyridinium Chlorochromate (PCC) in human urine.