Learn More
Norgen Biotek Corp Urine Cell-Free Circulating DNA Purification Maxi Kit
Catalog No. NC1753339
Encompass_Preferred
This kit provides a fast, reliable and convenient method to purify and concentrate high quality, high purity and inhibitor-free cell-free circulating DNA as well as viral DNA from fresh, preserved or frozen urine samples from volumes ranging from 10 mL to 30 mL. The first column will handle the large volume input of urine that is followed by a concentration on a mini column for a final elution of 50 µL to 100 µL. All components for the purification are provided in one convenient and fast kit for the easy processing of small input volumes of bodily fluids. For a schematic workflow of the protocol click here.The purified urine DNA is fully compatible with all downstream applications including PCR, qPCR, methylation-sensitive PCR and Southern Blot analysis, Microarrays and NGS.
Catalog No. NC1753339 Supplier Norgen Biotek Corp Supplier No. 56800
This kit provides a fast, reliable and convenient method to purify and concentrate high quality, high purity and inhibitor-free cell-free circulating DNA as well as viral DNA from fresh, preserved or frozen urine samples from volumes ranging from 10 mL to 30 mL. The first column will handle the large volume input of urine that is followed by a concentration on a mini column for a final elution of 50 µL to 100 µL. All components for the purification are provided in one convenient and fast kit for the easy processing of small input volumes of bodily fluids. For a schematic workflow of the protocol click here.The purified urine DNA is fully compatible with all downstream applications including PCR, qPCR, methylation-sensitive PCR and Southern Blot analysis, Microarrays and NGS.