Learn More
Norgen Biotek Corp Urine Collection and Preservation Tube 15 cc (50 Units)
Catalog No. NC1359874
Encompass_Preferred
Norgen's Urine Preservative is designed for the rapid preservation of DNA, RNA, microRNA and proteins from fresh urine samples. The Urine Preservative prevents the growth of Gram-negative and Gram-positive bacteria and fungi, and also inactivates viruses allowing the resulting non-infectious samples to be handled and shipped safely. In addition, the Urine Preservative eliminates the need to immediately process or freeze samples and allows the samples to be shipped to centralized testing facilities at ambient temperatures. The components of the Urine Preservative allow samples to be stored for over 2 years at room temperature with no detected degradation of urine DNA, RNA or proteins.
Catalog No. NC1359874 Supplier Norgen Biotek Corp Supplier No. 18122
Norgen's Urine Preservative is designed for the rapid preservation of DNA, RNA, microRNA and proteins from fresh urine samples. The Urine Preservative prevents the growth of Gram-negative and Gram-positive bacteria and fungi, and also inactivates viruses allowing the resulting non-infectious samples to be handled and shipped safely. In addition, the Urine Preservative eliminates the need to immediately process or freeze samples and allows the samples to be shipped to centralized testing facilities at ambient temperatures. The components of the Urine Preservative allow samples to be stored for over 2 years at room temperature with no detected degradation of urine DNA, RNA or proteins.