Learn More
Zymo Research Corporation Urine Collection Cup (120 ml), 10 pack
SDP
Catalog No. 502037035
Encompass_Preferred
Urine Conditioning Buffer effectively inhibits microbial growth in urine samples and the urine nucleic acid content is preserved at ambient temperature for up to 1 month. Urine Conditioning Buffer also facilitates pelleting of urine nucleic acids from large volume urine samples for the subsequent nucleic acids purification.
Catalog No. 50-203-7035 Supplier Zymo Research Corporation Supplier No. D3062
Urine Conditioning Buffer effectively inhibits microbial growth in urine samples and the urine nucleic acid content is preserved at ambient temperature for up to 1 month. Urine Conditioning Buffer also facilitates pelleting of urine nucleic acids from large volume urine samples for the subsequent nucleic acids purification.