Learn More
Alkali Scientific Urine Collection Cup with Inte
SDP
Supplier:  Alkali Scientific DTSC04
Encompass_Preferred
ASIUrine Collection and Direct Transfer Cups feature an integrated transfer mechanism designed for a closed system transfer of urine samples directly from the cup to a vacuum sample tube With the ASITransfer System laboratory personnel no longer need to open the collection cup and manually pour the sample into a tube Instead simply peel back the safety label and insert the vacuum tube for a safe efficient and reliable sample transfer Eliminates open sample transfer reducing spills improving lab hygiene and limiting exposure to hazardous samples Minimizes contamination risks for more accurate test results Directly transfers samples into vacuum tubes suitable for analysis and transport Compatible with most vacuum urine tubes Ensures consistent sample transfer volumes when used with compatible vacuum tubes Enhances efficiency and sample throughput compared to traditional open systems This system is designed to streamline laboratory workflows while prioritizing safety and reliability in
Catalog No. 50-302-7497