Learn More
G-Biosciences Urine Collection Kit
Catalog No. S04217
Change view
Click to view available options
No. per Pack:
10/Pk.
Catalog No. No. per Pack
S04217 10/Pk.
1 options
Catalog No. S04217 Supplier G-Biosciences Supplier No. 7862192
Add to Cart
Add to Cart
Urine collection kits are supplied with urine collection cups and urine conserve buffer.
Urine collection cups are used to transfer urine samples directly from the collection facilities to the testing laboratories. Urine Conserve Buffer is suitable for stabilizing DNA and RNA during storage and transport. It also preserves the urine by inhibiting microbial growth and is compatible to isolate DNA or RNA directly from urine sample without reagent removal.
Specifications
Product Type Urine Collection Kit
Includes Urine Collection Cups, Urine Conserve Buffer
No. per Pack 10/Pk.