Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marketlab, Inc. Urine Collection Labels

Catalog No. 22505479
Change view
Click to view available options
Length (English):
1.625 in.
1.75 in.
3.625 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
22-505-479 3.625 in.
22-505-449 1.625 in.
22-505-472 1.75 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-505-479 Supplier Marketlab, Inc. Supplier No. 6462
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Labels designed for urine collection identification and labeling needs.

  • Designed for use with urine collection labeling applications
  • Supports clear identification during urine collection processes
  • Supplied in blister packaging for convenient handling and storage

Specifications

Adhesive Type Peel and Stick
Coating None
Color Multi-colored
Finish Opaque
Format Label
Material Paper
Print Technology None
Product Type Label
Sterility Non-sterile
Length (English) 3.625 in.
Width (English) 3 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.