Learn More
Zymo Research Corporation Urine Conditioning Buffer (140 ml)
SDP
Catalog No. 501251365
Encompass_Preferred
The DNA/RNA Shield Urine Conditioning Buffer (UCB) allows sample storage and transport at ambient temperatures. There is no need for refrigeration or specialized equipment. With this product, the UCB can be added to any urine collection device.
Catalog No. 50-125-1365 Supplier Zymo Research Corporation Supplier No. D30611140
The DNA/RNA Shield Urine Conditioning Buffer (UCB) allows sample storage and transport at ambient temperatures. There is no need for refrigeration or specialized equipment. With this product, the UCB can be added to any urine collection device.