Learn More
Zymo Research Corporation Urine Conditioning Buffer (8 ml)
SDP
Catalog No. NC1805111
Encompass_Preferred
Features- Effectively preserves DNA and RNA in urine atambient temperatures- Facilitates pelleting of both cellular and cell-freenucleic acids from large volume urine samples- Inhibits microbial growth during long-term (cold-free)storage of urine samples
Catalog No. NC1805111 Supplier Zymo Research Corporation Supplier No. D306118
Features- Effectively preserves DNA and RNA in urine atambient temperatures- Facilitates pelleting of both cellular and cell-freenucleic acids from large volume urine samples- Inhibits microbial growth during long-term (cold-free)storage of urine samples