Learn More
G-Biosciences Urine Conserve Buffer
SCI_ED
Catalog No. S04216
Change view
Click to view available options
Volume (Metric):
100 mL
Catalog No. Volume (Metric)
S04216 100 mL
1 options
Catalog No. S04216 Supplier G-Biosciences Supplier No. 7862191
Add to Cart
Add to Cart
Urine Conserve Buffer preserves the urine by inhibiting microbial growth
Urine Conserve Buffer preserves the urine by inhibiting microbial growth. Urine Conserve Buffer is suitable for stabilizing DNA and RNA during storage and transport. It is compatible to isolate DNA or RNA directly from urine sample without reagent removal.
Specifications
Product Type Urine Collection Kit
Volume (Metric) 100 mL