Learn More
KB Med Urine Cup with Integrated Transfer Device
SDP
Catalog No. 21060056
Change view
Click to view available options
Sterility:
Sterile
Catalog No. Sterility
21-060-056 Sterile
1 options
Catalog No. 21-060-056 Supplier KB Med Supplier No. 61674
Add to Cart
Add to Cart
Sterile specimen cup with integrated transfer device designed for safe and convenient sample collection and handling.
  • Capacity: 120 mL
  • Yellow click top for secure closure
  • Integrated transfer device for easy sample handling
  • 250 per case
Specifications
Product Type Urine Collection System
Disposable Single-use
Graduated Yes
Material Polypropylene
Sterility Sterile
Color Clear
Latex Free Yes