Learn More
Bioworld Urine DNA Isolation Kit, 50 Prep(s)
Catalog No. 50196959
Encompass_Preferred
Fast and reliable purification of genomic and apoptotic DNA from urine Rapid isolation of small and large species of DNA from urine Convenient spin column format Purified DNA is highly suited to sensitive downstream applications FEATURES AND BENEFITSFast processing - Rapid spin-column format allows for the processing of multiple samples in 30 minutes. Isolation of both types of urine DNA - Isolate both high molecular weight DNA (greater than 1 kb in size; mostly cell associated) and the smaller DNA (150 - 250 bp; derived from the circulation). High quality DNA - Removal of highly concentrated salts, metabolic wastes and proteins provides high quality, concentrated DNA to be used in various downstream applications. Recovered DNA is suitable for downstream applications - Purified DNA is fully compatible with PCR, q-PCR and DNA fingerprinting
Catalog No. 50-196-959 Supplier Bioworld Supplier No. 107601021
Fast and reliable purification of genomic and apoptotic DNA from urine Rapid isolation of small and large species of DNA from urine Convenient spin column format Purified DNA is highly suited to sensitive downstream applications FEATURES AND BENEFITSFast processing - Rapid spin-column format allows for the processing of multiple samples in 30 minutes. Isolation of both types of urine DNA - Isolate both high molecular weight DNA (greater than 1 kb in size; mostly cell associated) and the smaller DNA (150 - 250 bp; derived from the circulation). High quality DNA - Removal of highly concentrated salts, metabolic wastes and proteins provides high quality, concentrated DNA to be used in various downstream applications. Recovered DNA is suitable for downstream applications - Purified DNA is fully compatible with PCR, q-PCR and DNA fingerprinting