Learn More
Bioworld Urine DNA/Protein Isolation Kit, 25 Prep(s)
Supplier:  Bioworld 107600861
Encompass_Preferred
For sequential isolation of total DNA and proteins from a single urine sample Isolate total urine DNA including exfoliated cell genomic DNA, apoptotic DNA and urine bacterial DNA Concentrate and desalt urinary proteins Versatile urine input amounts Rapid spin column format FEATURES AND BENEFITSIsolate a diversity of DNA species - The kit allows for the isolation of all DNA species found in urine, including exfoliated cell genomic DNA, apoptotic DNA and urine bacterial DNA Fast and easy processing - Rapid spin-column format allows for the processing of 12 samples in 15 minutes. Versatile urine input volume - Isolate urinary DNA and urinary proteins from 1 - 3 mL of urine samples. Isolate inhibitor-free DNA - Purified urinary DNA can be used in a number of sensitive downstream applications including PCR, q-PCR, Southern blot and Methylation sensitive PCR. Concentrate and desalt urinary proteins - Simultaneously concentrate and desalt up to 200 µg of urinary proteins
Catalog No. 50-196-943