Learn More
SA Scientific Urine hCG, Serum/Urine hCG, and Ultra™ hCG Serum/Urine Tests
Catalog No. SACOL087750
Change view
Click to view available options
Quantity:
3 x 5 mL
30 Tests
50 Tests
Type:
Serum Control Set
Serum, Urine
Ultra Serum Control Set
Ultra Serum/Urine Test
hCG Urine Control
hCG Urine Test
Sample Type:
Serum
Serum, Urine
Urine
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type Sample Type
SACOL087750 50 Tests Serum, Urine Serum, Urine
SACOL087731 30 Tests Ultra Serum/Urine Test Serum, Urine
SACOL087751 50 Tests Ultra Serum/Urine Test Serum, Urine
SACOL087712 3 x 5 mL Serum Control Set Serum
SACOL087715 3 x 5 mL Ultra Serum Control Set Serum
SACOL087413 3 x 5 mL hCG Urine Control Urine
SACOL087430 30 Tests hCG Urine Test Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. SACOL087750 Supplier SA Scientific Supplier No. COL087750
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Results in as little as four minutes
  • CLIA Complexity: Waived for Urine Only
  • CPT Codes: 81025 (Waived), 84703 (Non-waived)
  • Available with choice of sensitivity—10 or 20mIU/mL in Ultra Serum/Urine or 25mIU/mL in Serum/Urine and urine only tests
  • Visual, one-step test for the qualitative determination of human chorionic gonadotropin (hCG) in serum or urine to aid in early detection of pregnancy
  • Results in four to eight minutes
  • Utilizes combination of monoclonal and polyclonal antibody reagent to selectively detect elevated levels of hCG
  • Positive specimens react with the specific antibody-hCG colored conjugate and form a colored line in the S (specimen) area; absence of this colored line suggests a negative result
  • A colored line in the C (control) area will always appear regardless of the presence or absence of hCG
  • Addition of LH (300mIU/mL), FSH (1000mIU/mL) and TSH (1000μIU/mL) to negative (0mIU/mL hCG) or positive (25mIU/mL hCG) serum/urine samples showed no cross-reactivity
Specifications
Detectable Analytes hCG
Sample Type Serum, Urine
Type Serum, Urine
Quantity 50 Tests
Test Time 4 to 8 min.