Learn More
Norgen Biotek Corp Urine microRNA Purification Kit, purifies small RNA molecules (miRNA, tRNA, 5S rRNA), 0.5 - 1 mL sample input volume, 25 preps per kit, < 30 min processing time, spin column method
Catalog No. NC1906600
Encompass_Preferred
Urine microRNA Purification Kit, purifies small RNA molecules (miRNA, tRNA, 5S rRNA), 0.5 - 1 mL sample input volume, 25 preps per kit, < 30 min processing time, spin column method
Catalog No. NC1906600 Supplier Norgen Biotek Corp Supplier No. 29000
Urine microRNA Purification Kit, purifies small RNA molecules (miRNA, tRNA, 5S rRNA), 0.5 - 1 mL sample input volume, 25 preps per kit, < 30 min processing time, spin column method