Learn More
Bioworld Urine microRNA Purification Kit, 25 Prep(s)
Catalog No. 50196922
Encompass_Preferred
Rapid purification of microRNA from urine without phenol Purification of all small RNA molecules without the use of phenol Rapid and convenient spin-column protocol Isolate inhibitor-free urinary microRNAFEATURES AND BENEFITSFast and easy processing - Rapid spin-column format allows for the processing of multiple samples in under 30 minutes. Versatile urine input volume - Isolate urine microRNA from 0.5 - 3 mL of urine samples. No phenol:chloroform extractions - Norgen’s Urine microRNA Purification Kit isolates RNA without the use of harmful chemicals such as phenol or chloroform. Isolate all small RNA molecules – All small RNA species can be isolated including miRNA, siRNA, tRNA, and 5S rRNA. Isolate inhibitor-free RNA suitable for downstream applications - Purified RNA is fully compatible with various downstream applications relating to gene regulation and functional analysis, including RT-PCR, northern blotting and microarray analysis.
Catalog No. 50-196-922 Supplier Bioworld Supplier No. 107600671
Rapid purification of microRNA from urine without phenol Purification of all small RNA molecules without the use of phenol Rapid and convenient spin-column protocol Isolate inhibitor-free urinary microRNAFEATURES AND BENEFITSFast and easy processing - Rapid spin-column format allows for the processing of multiple samples in under 30 minutes. Versatile urine input volume - Isolate urine microRNA from 0.5 - 3 mL of urine samples. No phenol:chloroform extractions - Norgen’s Urine microRNA Purification Kit isolates RNA without the use of harmful chemicals such as phenol or chloroform. Isolate all small RNA molecules – All small RNA species can be isolated including miRNA, siRNA, tRNA, and 5S rRNA. Isolate inhibitor-free RNA suitable for downstream applications - Purified RNA is fully compatible with various downstream applications relating to gene regulation and functional analysis, including RT-PCR, northern blotting and microarray analysis.