Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals Urine Preservation Solution

Catalog No. MP116594350 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
MP116594350 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116594350 Supplier MP Biomedicals Supplier No. 116594350
Add to Cart
Add to Cart

Stabilizes cell-free DNA, genomic DNA, and nucleated cells in urine samples for up to 30 days when stored at room temperature.

  • Provides concurrent preservation of cell-free DNA, genomic DNA, and intact cells in urine samples, maintaining sample integrity during extended ambient storage.
  • Ensures reliable stabilization for periods of up to 30 days without the need for refrigeration, supporting flexible sample handling and transport.
  • Can be added directly to freshly collected urine samples, removing the requirement for centrifugation or prior separation of nucleated cells and cell-free DNA.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Urine Preservative Tube
Disposable Single-use
Graduated Yes
Material Polypropylene
Sterility Sterile
Color Yellow
Latex Free Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.