Learn More
JANT PHARMACAL CORPORATION ACCUSTRIP Urine Reagent Strip Reader for use with ACCUSTRIP URS 10 Urine Reagent Strips. *CLIA-Waived*
SDP
Catalog No. NC3457046
Encompass_Preferred
​The Accustrip URS Reader is a reflection photometer for the analysis of the Accustrip URS-10 urine test strips. When measurements are performed under standardized conditions, measured values may be displayed, printed and transferred to a computer. The instrument is designed for in-vitro diagnostic use (IVD) and should be used by healthcare professionals. 
Catalog No. NC3457046 Supplier JANT PHARMACAL CORPORATION Supplier No. UA850
​The Accustrip URS Reader is a reflection photometer for the analysis of the Accustrip URS-10 urine test strips. When measurements are performed under standardized conditions, measured values may be displayed, printed and transferred to a computer. The instrument is designed for in-vitro diagnostic use (IVD) and should be used by healthcare professionals. 