Learn More
Greiner Bio-One Urine Set
Catalog No. 22040555
Change view
Click to view available options
Bottom Shape:
Conical
Round
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bottom Shape
22-040-555 Conical
22-040-554 Round
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-040-555 Supplier Greiner Bio-One Supplier No. 453032
Add to Cart
Add to Cart
Urine Sets contain a VACUETTE™ urine tube and urine transfer unit.
  • Additive: CCM (Count and Culture Mannitol) additive
  • Tubes contain a stabilizer in powder form, which keeps the bacterial count in the sample stable at room temperature (20-25° C) for up to 48 hours
  • The good solubility of the additive stabilizes the urine sample quickly and is therefore especially suitable in cases when the analysis takes place later
  • This tube can be used as collection and/or transport container 
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Yellow with Black Ring
Sterility Non-sterile
Bottom Shape Conical
Description Single-packed; Non-ridged tube; 16 x 100
No. per Pack 300 Pk.
Volume (Metric) 4 mL
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov