Learn More
Pickering Laboratories Inc Artificial Urine, pH 6.5, Stabilized, each (200 mL/bottle)
SDP
Catalog No. 501354832
Encompass_Preferred
Our Artificial Urine solutions are formulated both according to official testing methods as well as published research on the use of artificial urine for corrosion testing and related applications. The products do not come sterile but could be sterilized before use using filtration. This ready-to-use solution has the same composition as Artificial Urine Medium and closely resembles human urine. This formulation contains non-toxic preservative to avoid bacteria growth and can be stored at room temperature. The pH of Artificial Urine Stabilized is 6.5
Catalog No. 50-135-4832 Supplier Pickering Laboratories Inc Supplier No. 17000600
Our Artificial Urine solutions are formulated both according to official testing methods as well as published research on the use of artificial urine for corrosion testing and related applications. The products do not come sterile but could be sterilized before use using filtration. This ready-to-use solution has the same composition as Artificial Urine Medium and closely resembles human urine. This formulation contains non-toxic preservative to avoid bacteria growth and can be stored at room temperature. The pH of Artificial Urine Stabilized is 6.5