Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Urine System Accessories

Catalog No. 22040114
Change view
Click to view available options
Product Type:
Urine Specimen Container
Urine Transfer Device
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-040-114 Urine Transfer Device
22-040-128 Urine Specimen Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-040-114 Supplier Greiner Bio-One Supplier No. 450251
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For hygienic transfer of urine from collection devices

TRUSTED_SUSTAINABILITY

Specifications

Product Type Urine Transfer Device
Material PET
Certifications/Compliance CE
For Use With (Application) For hygienic transfer of Urine from collection devices
Includes 10 cm Straw
Storage Requirements 4°C to 25°C
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.