Learn More
Labcon North America UrineTime™ II 24 Hour Urine Collection System, Bulk Pack of 5 Kits
Catalog No. NC9372066
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Container; Urine; 24 Hour; Labcon; UrineTime™ II; 24 Hour urine collection and proportioning system; Automatically splits urine samples into two equal containers; Allows for faster diagnosis; Perfect for trials and other clinical use; Each assembly kit Includes two polyethylene 1 liter containers, funnel, plugs, instructions, and disposal bags ; Not Autoclavable; Capacity: 1L; Includes 5 un-assembled kits per case; Made in USA
Catalog No. NC9372066 Supplier Labcon North America Supplier No. 40109200109
This item has a minimum qty of 2 per supplier requirements.
Container; Urine; 24 Hour; Labcon; UrineTime™ II; 24 Hour urine collection and proportioning system; Automatically splits urine samples into two equal containers; Allows for faster diagnosis; Perfect for trials and other clinical use; Each assembly kit Includes two polyethylene 1 liter containers, funnel, plugs, instructions, and disposal bags ; Not Autoclavable; Capacity: 1L; Includes 5 un-assembled kits per case; Made in USA