Learn More
BTNX Inc Qualitative Urine Toxicology Control - 12.26 (1neg/1pos control)
Catalog No. NC3361271
Encompass_Preferred
The Rapid Response™ Qualitative Urine Toxicology Control is packaged in amber vials (borosilicate glass) with silicone septas. The matrix is synthetic urine. This product lasts until the expiry date listed on the bottle, or 31 days after opening. For forensic use only. (12.26)
This item is not returnable. View return policy
Catalog No. NC3361271 Supplier BTNX Inc Supplier No. RR1VPN12.26
The Rapid Response™ Qualitative Urine Toxicology Control is packaged in amber vials (borosilicate glass) with silicone septas. The matrix is synthetic urine. This product lasts until the expiry date listed on the bottle, or 31 days after opening. For forensic use only. (12.26)