Learn More
Cliawaived Inc AMTOX UrineCheck 7 Controls
SDP
Catalog No. NC1859626
Encompass_Preferred
(3 x 25mL)Amtox UrineCheck Adulterant Controls. Refrigerated item for product stability.NOTE: This product is NOT CLIA waived. It is for FORENSIC USE ONLY and not for medical office use. Product should be protected from exposure to direct sunlight. The Amtox Controls are not for use in diagnosing disease or illness to effect treatment and/or cure.
This item is not returnable. View return policy
Catalog No. NC1859626 Supplier Cliawaived Inc Supplier No. AMTXURINECHECK7C
(3 x 25mL)Amtox UrineCheck Adulterant Controls. Refrigerated item for product stability.NOTE: This product is NOT CLIA waived. It is for FORENSIC USE ONLY and not for medical office use. Product should be protected from exposure to direct sunlight. The Amtox Controls are not for use in diagnosing disease or illness to effect treatment and/or cure.