Learn More
Teco Diagnostics UrineCheck 7 Strips, 25/pack
Catalog No. NC1422730
Encompass_Preferred
Teco UrineCheck 7 is a fast dip-and-read test for the determination of diluted or adulterated urine specimens. It is an important pre-screening test for any drug-testing program.
This item is not returnable. View return policy
Catalog No. NC1422730 Supplier Teco Diagnostics Supplier No. D70025
Teco UrineCheck 7 is a fast dip-and-read test for the determination of diluted or adulterated urine specimens. It is an important pre-screening test for any drug-testing program.