Learn More
Bioworld Urine (exfoliated cell) DNA Isolation Kit, 20 Prep(s)
Catalog No. 50196967
Encompass_Preferred
For the rapid and efficient purification of DNA from exfoliated cells in urine Isolate and detect genomic DNA from as little as 1 mL of urine High quality DNA for sensitive applications Rapid processing time from sampling to downstream testing. FEATURES AND BENEFITS: Fast processing - Rapid spin-column format allows for the processing of multiple samples in 45 minutes (including a 30 minute incubation). Isolate genomic DNA from small volumes of urine - DNA can be isolated and detected from the exfoliated cells found in as little as 1 mL of urine. High quality DNA - Removal of highly concentrated salts, metabolic wastes and proteins provides high quality DNA to be used in various downstream applications. Recovered DNA is suitable for downstream applications - Purified DNA is fully compatible with PCR, quantitative PCR and Southern Blot analysis.
Catalog No. 50-196-967 Supplier Bioworld Supplier No. 107601101
For the rapid and efficient purification of DNA from exfoliated cells in urine Isolate and detect genomic DNA from as little as 1 mL of urine High quality DNA for sensitive applications Rapid processing time from sampling to downstream testing. FEATURES AND BENEFITS: Fast processing - Rapid spin-column format allows for the processing of multiple samples in 45 minutes (including a 30 minute incubation). Isolate genomic DNA from small volumes of urine - DNA can be isolated and detected from the exfoliated cells found in as little as 1 mL of urine. High quality DNA - Removal of highly concentrated salts, metabolic wastes and proteins provides high quality DNA to be used in various downstream applications. Recovered DNA is suitable for downstream applications - Purified DNA is fully compatible with PCR, quantitative PCR and Southern Blot analysis.