Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copan Diagnostics UriSponge™ For Urine Collection, Transport, and Preservation System
SDP

Catalog No. 22040564 Shop All Copan Diagnostics Inc. Products
Change view
Click to view available options
Tube Size:
Total length: 81.30 mm ± 0.40 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tube Size
22-040-564 Total length: 81.30 mm ± 0.40 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-040-564 Supplier Copan Diagnostics Supplier No. 8U033S50
Add to Cart
Add to Cart
This item is not returnable. View return policy

UriSponge™ ensures reliable urine collection, transport and preservation of the specimen, while maintaining integrity for accurate uropathogenic bacteria and yeasts cultivation using standard clinical lab procedures. It is safe and reliable.

UriSponge™ is an innovative urine collection, transport, and preservation system designed to simplify workflows, enhance specimen integrity, and improve laboratory efficiency. UriSponge™ features a screw cap tube with a plastic applicator and two cylindrical sponges with preservatives. Upon contact with urine, Sodium Propionate and Potassium Sorbate, maintain sample stability throughout transport, providing dependable results and minimizing the risk of contamination or degradation. This design ensures efficient specimen transport from collection to laboratory while maintaining integrity for accurate uropathogenic bacteria and yeasts cultivation using standard clinical laboratory procedures.

  • No fill line necessary: UriSponge™ removes the need for precise fill lines, minimizing sample rejection and reducing repeat collection.
  • Safe and reliable: Eliminates the need for hypodermic needles and urine transfer straws needed, reducing the risk of accidental injury, and reducing waste.
  • Cost effective: Minimizes expenses and waste by eliminating consumables like needles and transfer straws, and preventing the need for repeat collections due to inaccurate filling.
  • Automation ready: Designed for seamless integration and automated specimen processors like WASP™. UriSponge™ enhances laboratory workflow by supporting standardized, high throughput sample processing, leading to greater consistency and efficiency.
  • New Preservatives: UriSponge™ has the following preservative substance (urine maintenance formula)
  • Boric acid free: Unique urine preservative formulation is non-hazardous and non-toxic.
  • FDA Cleared: Innovative Urine Collection Device received FDA clearance in October of 2024.

Efficient Handling and Processing

  • UriSponge™ enhances efficiency in handling and processing with its versatile and compact design. It supports both automated planting and streaking and manual centrifugation for plating, offering flexibility in laboratory workflows.

Seamless Automation Integration

  • UriSponge™ enhances the utility of Copan's WASP™ Walk-Away Specimen Processor for automating upfront microbiology specimen processing. With capabilities including planting and streaking, Gram slide preparation, and inoculation of enrichment broths, WASP™ redefines laboratory efficiency. UriSponge™ seamlessly integrates with this automated specimen processing, ensuring a smooth workflow from urine sample collection to processing.

Specifications

Product Type Urine Collection System
Closure Color Yellow
Closure Type Screw cap
Disposable Single-use
Graduated No
Material Polypropylene
Sterility Sterile
Additive Sodium Propionate and Potassium Sorbate
Bottom Shape Flat
Certifications/Compliance FDA cleared 10/24
Closure Material High Density Polyethylene
Closure Size 15.00 mm ± 0.20 mm
Color Clear
Diameter (Metric) Inner 11.50 mm ± 0.20 mm
Format Tube
For Use With (Application) Urine collection, transport, and preservation
For Use With (Equipment) Automation instruments such as Copan's WASP and WASPLab
Latex Free Yes
No. per Case 300
No. per Pack 50
Packaging Plastic Envelope
Storage Requirements 2-25C - DO NOT FREEZE
Tube Size Total length: 81.30 mm ± 0.40 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.