Learn More
CST TECHNOLOGIES UriSub, pH 7.4, 1000 mL
Catalog No. NC9591461
Encompass
UriSub is a liquid, particulate-free, odorless stable synthetic urine for use in manufacturing of urine controls and reference standards for clinical chemistry and immunological procedures.
Catalog No. NC9591461 Supplier CST TECHNOLOGIES Supplier No. US22051000
UriSub is a liquid, particulate-free, odorless stable synthetic urine for use in manufacturing of urine controls and reference standards for clinical chemistry and immunological procedures.