Learn More
Apexbio Technology LLC URMC-099, 100mg CAS# 1229582-33-5
SDP
Supplier:  Apexbio Technology LLC B4877100
Encompass_Preferred
URMC-099 is an orally bioavailable and brain penetrant inhibitor of MLK3 with IC50 value of 14 nM [1].Mixed Lineage Kinase 3 (MLK3) is a member of the serine/threonine kinase family that regulate MAPK8/JNK signaling cascade and also activates ERK and p38. MLK3 activation can result in the stimulation of cell motility and migration [1] [2].URMC-099 is a novel and orally bioavailable MLK3 inhibitor. In murine BV-2 microglial cells, URMC-099 inhibited LPS-induced TNFrelease. In primary human monocytes, URMC-099 inhibited HIV-1 Tat-induced release of cytokines [1]. In neutrophils, URMC-099 reduced chemotaxis induced by fMLP and inhibited fMLP-stimulated F-actin formation [2].In Tat-injected brains of mice, URMC-099 inhibited up-regulation of phospho-JNK. In HIV-1 Associated Neurocognitive Disorders (HAND) preclinical models, URMC-099 inhibited multiple kinase pathways, including MLK3 and LRRK2 (IC50 = 11 nM). Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8389