Learn More
Medchemexpress LLC HY-12599 10mg Medchemexpress, URMC-099 CAS:1229582-33-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1259910MG
Encompass_Preferred
Medchemexpress, HY-12599 10mg URMC-099 CAS:1229582-33-5 URMC-099 is an orally bioavailable and potent mixed lineage kinase type 3 (MLK3) (IC50=14 nM) inhibitor with with excellent blood-brain barrier penetration properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8897