Learn More
Selleck Chemical LLC URMC-099 5mg 1229582-33-5
SDP
Supplier:  Selleck Chemical LLC S73435MG
Encompass_Preferred
URMC-099 is an orally bioavailable, brain penetrant mixed lineage kinase (MLK) inhibitor with IC50 of 19 nM, 42 nM, 14 nM, and 150 nM, for MLK1, MLK2, MLK3, and DLK, respectively, and also inhibits LRRK2 activity with IC50 of 11 nM. URMC-099 also inhibits ABL1 with IC50 of 6.8 nM. URMC-099 induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4669