Learn More
Norgen Biotek Corp Urne Cellfree Circ Rna Minikit
Catalog No. NC1847984
Encompass_Preferred
This kit provides a fast, reliable and convenient method to purify and concentrate high quality, high purity and inhibitor-free cell-free circulating RNA, including exosomal RNA as well as viral RNA from fresh, preserved or frozen urine samples from volumes ranging from 250 µL to 2 mL. All components for the purification are provided in one convenient and fast kit for the easy processing of small input volumes of bodily fluids. The purified urine RNA is fully compatible with all downstream applications including PCR, qPCR, methylation-sensitive reverse transcription qPCR, reverse transcription PCR, Northern blotting, RNase protection and primer extension, expression array assays, and NGS.
Catalog No. NC1847984 Supplier Norgen Biotek Corp Supplier No. 56900
This kit provides a fast, reliable and convenient method to purify and concentrate high quality, high purity and inhibitor-free cell-free circulating RNA, including exosomal RNA as well as viral RNA from fresh, preserved or frozen urine samples from volumes ranging from 250 µL to 2 mL. All components for the purification are provided in one convenient and fast kit for the easy processing of small input volumes of bodily fluids. The purified urine RNA is fully compatible with all downstream applications including PCR, qPCR, methylation-sensitive reverse transcription qPCR, reverse transcription PCR, Northern blotting, RNase protection and primer extension, expression array assays, and NGS.