Learn More
Medchemexpress LLC Urocanic acid | 104-98-3 | 99.9% | 138.13 | 1 ML
SDP
Catalog No. 5000439706
Encompass_Preferred
Urocanic acid, produced in the upper layers of mammalian skin, is a major absorber of ultraviolet radiation (UVR). It is formed in the upper layers of the epidermis where filaggrin, a histidine-rich filamentous protein, is broken down by proteinases into component amino acids after caspase-14 cleavage of profilaggrin. It is for research use only.
  • Major absorber of ultraviolet radiation (UVR)
  • Endogenous metabolite
  • Disease markers
  • Human gut microbiota metabolites
  • Ketones, aldehydes, acids
Catalog No. 50-004-39706 Supplier Medchemexpress LLC Supplier No. HY11300810MM/1ML
Urocanic acid, produced in the upper layers of mammalian skin, is a major absorber of ultraviolet radiation (UVR). It is formed in the upper layers of the epidermis where filaggrin, a histidine-rich filamentous protein, is broken down by proteinases into component amino acids after caspase-14 cleavage of profilaggrin. It is for research use only.
  • Major absorber of ultraviolet radiation (UVR)
  • Endogenous metabolite
  • Disease markers
  • Human gut microbiota metabolites
  • Ketones, aldehydes, acids